Tisk…

2-Nitrobenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

N11507-100G N11507-25G
SIALN11507-100GEA 3080 CZK
SIALN11507-100G SIALN11507-25G
2-Nitrobenzenesulphonyl chloride, Sigma-Aldrich®
2-Nitrobenzenesulphonyl chloride
Vzorec: C₆H₄ClNO₄S
Storage Temperature: Ambient
MDL Number: MFCD00007430
CAS číslo: 1694-92-4
EINECS: 216-907-1
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR