Tisk…

1-Methyl-2-piperidone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

M73788-10G M73788-25G
SIALM73788-10GEA 5920 CZK
SIALM73788-10G SIALM73788-25G
1-Methyl-2-piperidone, Sigma-Aldrich®
1-Methyl-2-piperidone
Vzorec: C₆H₁₁NO
MDL Number: MFCD00006555
CAS číslo: 931-20-4
EINECS: 213-231-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR