Tisk…

Methyl cellulose, 25 mPa.S (2% H₂O), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cellulose methyl ether

M-Clarity™ quality level = MQ200

M6385-100G M6385-250G M6385-500G
SIALM6385-100GEA 3850 CZK
SIALM6385-100G SIALM6385-250G SIALM6385-500G
Methyl cellulose, 25 mPa.S (2% H₂O), Sigma-Aldrich®
Methyl cellulose

1 mPa.s = 1cP

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS číslo: 9004-67-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR