Tisk…

Methylakrylát stabilised, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acrylic acid methyl ester

M-Clarity™ quality level = MQ200

M27301-1L M27301-18L M27301-250ML M27301-5ML
SIALM27301-1LEA 2150 CZK
SIALM27301-1L SIALM27301-18L SIALM27301-250ML SIALM27301-5ML
Methylakrylát stabilised, Sigma-Aldrich®
Methylakrylát
Vzorec: C₄H₆O₂
MDL Number: MFCD00008627
CAS číslo: 96-33-3
EINECS: 202-500-6
UN: 1919
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR