Tisk…

4-Methoxycinnamic acid predominantly trans 99%, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 3-(4-Methoxyphenyl)acrylic acid

M-Clarity™ quality level = MQ100

M13807-100G
SIALM13807-100GEA 9670 CZK
SIALM13807-100G
4-Methoxycinnamic acid predominantly trans 99%, Sigma-Aldrich®
4-Methoxycinnamic acid
Vzorec: C₁₀H₁₀O₃
MW: 178.18 g/mol
MDL Number: MFCD00004398
CAS číslo: 830-09-1
EINECS: 212-594-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR