Tisk…

Methyl cellulose, 1500 mPa.S (2% H₂O), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cellulose methyl ether

M-Clarity™ quality level = MQ200

M0387-100G M0387-1KG M0387-500G
SIALM0387-100GEA 3640 CZK
SIALM0387-100G SIALM0387-1KG SIALM0387-500G
Methyl cellulose, 1500 mPa.S (2% H₂O), Sigma-Aldrich®
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS číslo: 9004-67-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR