Tisk…

L(+)-Lyzin monohydrochlorid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

L8662-100G L8662-1KG L8662-25G
SIALL8662-100GEA 3260 CZK
SIALL8662-100G SIALL8662-1KG SIALL8662-25G
L(+)-Lyzin monohydrochlorid, Sigma-Aldrich®
L(+)-Lyzin monohydrochlorid
Vzorec: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Bod tání: 263…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS číslo: 657-27-2
EINECS: 211-519-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR