Tisk…

Lactic acid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

L6661-100ML L6661-500ML
SIALL6661-100MLEA 5440 CZK
SIALL6661-100ML SIALL6661-500ML
Lactic acid, Sigma-Aldrich®
Lactic acid
Vzorec: H₃CCH(OH)COOH
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS číslo: 50-21-5
EINECS: 200-018-0
Merck Index: 13,05351

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR