Tisk…

Lidocaine, Sigma-Aldrich®

Dodavatel: Merck


MQ200

L1026-1VL
SIALL1026-1VLEA 3780 CZK
SIALL1026-1VL
Lidocaine, Sigma-Aldrich®
Lidocaine
Vzorec: C₁₄H₂₂N₂O
MW: 234.34
MDL Number: MFCD00026733
CAS číslo: 137-58-6
EINECS: 205-302-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR