Tisk…

Kanamycin sulphate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ300

K1637-1G K1637-25G K1637-5G
SIALK1637-1GEA 4700 CZK
SIALK1637-1G SIALK1637-25G SIALK1637-5G
Kanamycin sulphate, Sigma-Aldrich®
Kanamycin sulphate
Vzorec: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS číslo: 25389-94-0
EINECS: 246-933-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR