Tisk…

2-(4-Hydroxyphenylazo)benzoic acid, Sigma-Aldrich®

Dodavatel: Merck

H5126-1G H5126-25G H5126-5G
SIALH5126-1GEA 489 CZK
SIALH5126-1G SIALH5126-25G SIALH5126-5G
2-(4-Hydroxyphenylazo)benzoic acid, Sigma-Aldrich®
2-(4-Hydroxyphenylazo)benzoic acid
Vzorec: C₁₃H₁₀N₂O₃
MDL Number: MFCD00002428
CAS číslo: 1634-82-8
EINECS: 216-655-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR