Tisk…

4-Hydroxybenzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol

M-Clarity™ quality level = MQ200

H20806-100G H20806-10G H20806-25G
SIALH20806-100GEA 7350 CZK
SIALH20806-100G SIALH20806-10G SIALH20806-25G
4-Hydroxybenzyl alcohol, Sigma-Aldrich®
4-Hydroxybenzyl alcohol
Vzorec: C₇H₈O₂
MW: 124.14
Bod varu: 252
Bod tání: 115…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004658
CAS číslo: 623-05-2
EINECS: 210-768-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR