Tisk…

Glycine methyl ester hydrochloride (H-Gly-OMe.HCl), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

G6600-100G G6600-500G G6600-5G
SIALG6600-100GEA 1310 CZK
SIALG6600-100G SIALG6600-500G SIALG6600-5G
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl), Sigma-Aldrich®
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)
Vzorec: C₃H₈ClNO₂
MDL Number: MFCD00012870
CAS číslo: 5680-79-5
EINECS: 227-139-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR