Tisk…

Glycerol phosphate disodium salt hydrate mixture of isomers, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

G6501-100G G6501-1KG G6501-25G G6501-500G
SIALG6501-100GEA 5150 CZK
SIALG6501-100G SIALG6501-1KG SIALG6501-25G SIALG6501-500G
Glycerol phosphate disodium salt hydrate mixture of isomers, Sigma-Aldrich®
Glycerol phosphate disodium salt hydrate

Glycerol phosphate disodium salt was used in a novel microplate-based chromatin immunoprecipitation method to immunoprecipitate DNA from rat mesangial cells in a 96-well format.


Usually contains 50% β-isomer and 50% rac-α-isomer

Vzorec: C₃H₇Na₂O₆P·H₂O
MDL Number: MFCD00149082
CAS číslo: 55073-41-1
EINECS: 212-464-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR