Tisk…
Gel filtration media, Sephadex® G-50
Dodavatel: Merck
G50150-10G G50150-50G G5050-100G G5050-10G G5050-50G G5080-10G G5080-50G
SIALG50150-10GEA 4920 CZK
SIALG50150-10G SUPLG50150-50G SIALG5050-100G SIALG5050-10G SIALG5050-50G SIALG5080-10G SIALG5080-50G
Gel filtration media, Sephadex® G-50
Systémy pro purifikaci proteinů Protein Purification Resins
Sephadex® G-50 is a gel filtration medium used in affinity chromatography, protein chromatrography and gel filtration chromatography.
  • Swelling from 1 g to (9 to 11 ml)
It has been used to investigate effects from snake venom, isolate lectins from various species and to demonstrate the positive influence of 20% ethanol on proteinase desorption.
Balení: Packed in poly bottle.