Tisk…

EDTA trisodium salt monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate, Trisodium hydrogen ethylenediaminetetraacetate monohydrate, Ethylenediaminetetra-acetic acid trisodium salt monohydrate, Ethylenedinitrilotetra-acetic acid trisodium salt monohydrate, Edetic acid trisodium salt monohydrate

M-Clarity™ quality level = MQ200

ED3SS-100G ED3SS-1KG ED3SS-500G
SIALED3SS-100GEA 3640 CZK
SIALED3SS-100G SIALED3SS-1KG SIALED3SS-500G
EDTA trisodium salt monohydrate, Sigma-Aldrich®
EDTA trisodium salt monohydrate
Vzorec: C₁₀H₁₃N₂Na₃O₈·1H₂O
MW: 376.21
Storage Temperature: Ambient
CAS číslo: 10378-22-0
EINECS: 205-758-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR