Tisk…

3-Methylcrotonic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

D138606-100G D138606-500G
SIALD138606-100GEA 2230 CZK
SIALD138606-100G SIALD138606-500G
3-Methylcrotonic acid, Sigma-Aldrich®
3-Methylcrotonic acid
Vzorec: C₅H₈O₂
MDL Number: MFCD00004366
CAS číslo: 541-47-9
EINECS: 208-782-7
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR