Tisk…

Galactitol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol

M-Clarity™ quality level = MQ200

D0256-100G
SIALD0256-100GEA 4050 CZK
SIALD0256-100G
Galactitol, Sigma-Aldrich®
Galactitol
Vzorec: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17
Storage Temperature: Ambient
MDL Number: MFCD00064288
CAS číslo: 608-66-2
EINECS: 210-165-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR