Tisk…

Cholesterol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

M-Clarity™ quality level = MQ200

C8667-500MG C8667-100G C8667-1G C8667-25G C8667-5G
SIALC8667-500MGEA 1660 CZK
SIALC8667-500MG SIALC8667-100G SIALC8667-1G SIALC8667-25G SIALC8667-5G
Cholesterol, Sigma-Aldrich®
Cholesterol
Vzorec: C₂₇H₄₆O
MW: 386.66
Bod varu: 360
Bod tání: 147…150 °C
Hustota: 1.07 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00003646
CAS číslo: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR