Tisk…

CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

C2154-100G C2154-1KG
SIALC2154-100GEA 3940 CZK
SIALC2154-100G SIALC2154-1KG
CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt), Sigma-Aldrich®
CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt)
Vzorec: C₉H₁₈NNaO₄S
MDL Number: MFCD00070063
CAS číslo: 102601-34-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR