Tisk…

Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2), Sigma-Aldrich®

Dodavatel: Merck

C1506-100MG C1506-10MG C1506-250MG C1506-25MG C1506-1G
SIALC1506-100MGEA 3080 CZK
SIALC1506-100MG SIALC1506-10MG SIALC1506-250MG SIALC1506-25MG SIALC1506-1G
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2), Sigma-Aldrich®
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2)
Vzorec: C₉H₁₄N₃Na₂O₁₄P₃
MDL Number: MFCD00078193
CAS číslo: 36051-68-0
EINECS: 252-849-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR