Tisk…

L(-)-Carnitine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

C0158-1G C0158-100G C0158-25G C0158-5G
SIALC0158-1GEA 1560 CZK
SIALC0158-1G SIALC0158-100G SIALC0158-25G SIALC0158-5G
L(-)-Carnitine, Sigma-Aldrich®
L(-)-Carnitine
Vzorec: C₇H₁₅NO₃
MDL Number: MFCD00038747
CAS číslo: 541-15-1
EINECS: 208-768-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR