Tisk…

zinek

Dodavatel: Merck

BCR327-1EA
SIALBCR327-1EAEA 13260 CZK
SIALBCR327-1EA
zinek
zinek
Vzorec: Zn
MW: 65.39
Bod varu: 908
Bod tání: 420
Hustota: 7.14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS číslo: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR