Tisk…

α-Bromo-o-xylene, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 2-Methylbenzyl bromide, alpha-Bromo-o-xylene

M-Clarity™ quality level = MQ100

B83401-25G
SIALB83401-25GEA 1870 CZK
SIALB83401-25G
α-Bromo-o-xylene, Sigma-Aldrich®
α-Bromo-o-xylene
Vzorec: C₈H₉Br
Bod varu: 216 °C (1013 hPa)
Bod tání: 18…20 °C
Hustota: 1,381 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000174
CAS číslo: 89-92-9
EINECS: 201-951-6
UN: 1701
ADR: 6.1,II
Merck Index: 12,10223

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR