Tisk…

(±)-α-Bromo-γ-butyrolactone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

B59608-100G B59608-25G
SIALB59608-100GEA 4890 CZK
SIALB59608-100G SIALB59608-25G
(±)-α-Bromo-γ-butyrolactone, Sigma-Aldrich®
(±)-α-Bromo-γ-butyrolactone
Vzorec: C₄H₅BrO₂
Bod varu: 137…139 °C (8 hPa)
Hustota: 1,994 g/cm³ (20 °C)
MDL Number: MFCD00005387
CAS číslo: 5061-21-2
EINECS: 225-764-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR