Tisk…

5-Bromo-2'-deoxyuridine (broxuridine), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

B5002-100G B5002-100MG B5002-1G B5002-250MG B5002-500MG B5002-5G
SIALB5002-100GEA 0 CZK
SIALB5002-100G SIALB5002-100MG SIALB5002-1G SIALB5002-250MG SIALB5002-500MG SIALB5002-5G
5-Bromo-2'-deoxyuridine (broxuridine), Sigma-Aldrich®
5-Bromo-2'-deoxyuridine (broxuridine)
Vzorec: C₉H₁₁BrN₂O₅
MDL Number: MFCD00006529
CAS číslo: 59-14-3
EINECS: 200-415-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR