Tisk…

2,2'-Dihydroxydiphenylmethane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Bis(o-hydroxyphenyl)methane, 2,2'-Methylenediphenol

M-Clarity™ quality level = MQ100

B46808-1G
SIALB46808-1GEA 2250 CZK
SIALB46808-1G
2,2'-Dihydroxydiphenylmethane, Sigma-Aldrich®
2,2'-Dihydroxydiphenylmethane
Vzorec: C₁₃H₁₂O₂
MDL Number: MFCD00002243
CAS číslo: 2467-02-9
EINECS: 219-578-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR