Tisk…

Adenine sulphate, Sigma-Aldrich®

Dodavatel: Merck

A9126-100G A9126-25G A9126-1KG
SIALA9126-100GEA 14010 CZK
SIALA9126-100G SIALA9126-25G SIALA9126-1KG
Adenine sulphate, Sigma-Aldrich®
Adenine sulphate
Vzorec: C₅H₅N₅·½H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS číslo: 321-30-2
EINECS: 206-286-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR