Tisk…

Kyselina L(+)-asparágová, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D

M-Clarity™ quality level = MQ200

A8949-25G
SIALA8949-25GEA 1440 CZK
SIALA8949-25G
Kyselina L(+)-asparágová, Sigma-Aldrich®
Kyselina L(+)-asparágová
Vzorec: C₄H₇NO₄
MW: 133.1
Bod varu: 311
Bod tání: 251
Hustota: 1.6613
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS číslo: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR