Tisk…

2-Aminobenzenesulphonic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

A86805-100G
SIALA86805-100GEA 1750 CZK
SIALA86805-100G
2-Aminobenzenesulphonic acid, Sigma-Aldrich®
2-Aminobenzenesulphonic acid
Vzorec: C₆H₇NO₃S
MDL Number: MFCD00007705
CAS číslo: 88-21-1
EINECS: 201-810-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR