Tisk…

N-Acetyl-β-D-glucosamine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

A8625-10MG A8625-5G
SIALA8625-10MGEA 1400 CZK
SIALA8625-10MG SIALA8625-5G
N-Acetyl-β-D-glucosamine, Sigma-Aldrich®
N-Acetyl-β-D-glucosamine
Vzorec: C₈H₁₅NO₆
Storage Temperature: Ambient
MDL Number: MFCD00136044
CAS číslo: 7512-17-6
EINECS: 231-368-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR