Tisk…

L(+)-Arginin 98.5-101.0% Ph. Eur., USP cell culture reagent 98.5-101.0%, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

M-Clarity™ quality level = MQ200

A8094-100G A8094-10MG A8094-1KG A8094-25G
SIALA8094-100GEA 4330 CZK
SIALA8094-100G SIALA8094-10MG SIALA8094-1KG SIALA8094-25G
L(+)-Arginin 98.5-101.0% Ph. Eur., USP cell culture reagent 98.5-101.0%, Sigma-Aldrich®
L(+)-Arginin
Vzorec: C₆H₁₄N₄O₂
Bod varu: 368 °C (1013 hPa)
Bod tání: 238 °C
Hustota: 1,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS číslo: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785

Objednat

Výsledky specifikačního testu

Appearance (Color) White
Appearance (Form) Crystalline Powder
Solubility (Color) Colorless
Solubility (Turbidity) at 1.0 g to 10 ml of H2O Clear
Endotoxin Assay < 1 EU/mg
Identity by IR Conforms to Structure
Origin (Must Be Non-Animal Source) Non-Animal Source
Specific Rotation C = 8 in 6 M HCl (20 Deg C/EP) 26.9 - 27.9 °
Specific Rotation C = 8 in 6 M HCl (25 Deg C/USP) 26.3 - 27.7 °
Ammonium <= 0.02%
Sulfate <= 0.020%
Iron (Fe) <= 10 ppm
Chloride <= 0.020%
Loss on Drying < 0.50 %
Residue on ignition (Ash) < 0.10 %
Ninhydrin-Positive Substances (Other amino acids) Conforms
pH (5% in H2O) 10.5 - 12.0
Assay (Dry Basis) 98.5 - 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR