Tisk…

N(α)-Acetyl-L-tryptophan, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

A6376-10G A6376-25G
SIALA6376-10GEA 2380 CZK
SIALA6376-10G SIALA6376-25G
N(α)-Acetyl-L-tryptophan, Sigma-Aldrich®
N(α)-Acetyl-L-tryptophan
Vzorec: C₁₃H₁₄N₂O₃
MDL Number: MFCD00065976
CAS číslo: 1218-34-4
EINECS: 214-935-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR