Tisk…

3-Amino-2-chloropyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

A46900-10G A46900-50G
SIALA46900-10GEA 1070 CZK
SIALA46900-10G SIALA46900-50G
3-Amino-2-chloropyridine, Sigma-Aldrich®
3-Amino-2-chloropyridine
Vzorec: C₅H₅ClN₂
Bod varu: 263 °C (1013 hPa)
Bod tání: 78…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006238
CAS číslo: 6298-19-7
EINECS: 228-572-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR