Tisk…

Ampicillin sodium salt, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

M-Clarity™ quality level = MQ200

A0166-100G A0166-25G A0166-5G
SIALA0166-100GEA 88860 CZK
SIALA0166-100G SIALA0166-25G SIALA0166-5G
Ampicillin sodium salt, Sigma-Aldrich®
Ampicillin sodium salt
Vzorec: C₁₆H₁₈N₃NaO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS číslo: 69-52-3
EINECS: 200-708-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR