Tisk…

Vitamin K1 (phytomenadione, phylloquinone), Sigma-Aldrich®

Dodavatel: Merck

95271-250MG 95271-1G
SIAL95271-250MGEA 1110 CZK
SIAL95271-250MG SIAL95271-1G
Vitamin K1 (phytomenadione, phylloquinone), Sigma-Aldrich®
Vitamin K1 (phytomenadione, phylloquinone)
Vzorec: C₃₁H₄₆O₂
MDL Number: MFCD00214063
CAS číslo: 84-80-0
EINECS: 201-564-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR