Tisk…

Triethylenetetramine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: N,N'-Bis(2-aminoethyl)-1,2-ethanediamine, Trientine, DEH 24, EPH 925

90460-10ML 90460-50ML
SIAL90460-10MLEA 4070 CZK
SIAL90460-10ML SIAL90460-50ML
Triethylenetetramine, Sigma-Aldrich®
Triethylenetetramine
Vzorec: C₆H₁₈N₄
MW: 146.24
MDL Number: MFCD00008169
CAS číslo: 112-24-3
EINECS: 203-950-6
UN: 2259
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR