Tisk…

Semikarbazid hydrochlorid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride

M-Clarity™ quality level = MQ200

84940-100G
SIAL84940-100GEA 0 CZK
SIAL84940-100G
Semikarbazid hydrochlorid, Sigma-Aldrich®
Semikarbazid hydrochlorid
Vzorec: CH₆ClN₃O
Bod tání: 174…178 °C
MDL Number: MFCD00013009
CAS číslo: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,08588

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR