Tisk…

Phenylglyoxylic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

78610-10G
SIAL78610-10GEA 2090 CZK
SIAL78610-10G
Phenylglyoxylic acid, Sigma-Aldrich®
Phenylglyoxylic acid
Vzorec: C₈H₆O₃
MDL Number: MFCD00002575
CAS číslo: 611-73-4
EINECS: 210-278-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR