Tisk…

1-Butyl-3-methylimidazolium octylsulphate, Sigma-Aldrich®

Dodavatel: Merck

75059-50G-F 75059-5G-F
SIAL75059-50G-FEA 8660 CZK
SIAL75059-50G-F SIAL75059-5G-F
1-Butyl-3-methylimidazolium octylsulphate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium octylsulphate
Vzorec: C₁₆H₃₂N₂O₄S
MDL Number: MFCD03788914
CAS číslo: 445473-58-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR