Tisk…

Nordihydroguaiaretic acid, Sigma-Aldrich®

Dodavatel: Merck

74540-1G 74540-5G
SIAL74540-1GEA 2630 CZK
SIAL74540-1G SIAL74540-5G
Nordihydroguaiaretic acid, Sigma-Aldrich®
Nordihydroguaiaretic acid
Vzorec: C₁₈H₂₂O₄
MDL Number: MFCD00002206
CAS číslo: 500-38-9
EINECS: 207-903-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR