Tisk…

Bis(tributyltin)sulphide, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

720585-1G
SIAL720585-1GEA 1900 CZK
SIAL720585-1G
Bis(tributyltin)sulphide, Sigma-Aldrich®
Bis(tributyltin)sulphide
Vzorec: C₂₄H₅₄SSn₂
MDL Number: MFCD00048750
CAS číslo: 4808-30-4
EINECS: 225-369-7
UN: 2788
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR