Tisk…

Tributyl(pentafluoroethyl)stannane, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

711063-1G
SIAL711063-1GEA 3090 CZK
SIAL711063-1G
Tributyl(pentafluoroethyl)stannane, Sigma-Aldrich®
Tributyl(pentafluoroethyl)stannane
Vzorec: C₁₄H₂₇F₅Sn
MDL Number: MFCD04116432
CAS číslo: 1426-66-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR