Tisk…

Cyclobutylboronic acid MIDA ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 8-Cyclobutyl-4-methyl-2,6-dioxohexahydro-[1,3,2]oxazaborolo[2,3-b][1,3,2]oxazaborol-4-ium-8-uide

710040-1G
SIAL710040-1GEA 3210 CZK
SIAL710040-1G
Cyclobutylboronic acid MIDA ester, Sigma-Aldrich®
Cyclobutylboronic acid MIDA ester
Vzorec: C₉H₁₄BNO₄
MW: 211.0228 g/mol
MDL Number: MFCD15144797
CAS číslo: 1104637-37-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR