Tisk…

4-Iodophenylboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

699470-1G 699470-5G
SIAL699470-1GEA 2460 CZK
SIAL699470-1G SIAL699470-5G
4-Iodophenylboronic acid pinacol ester, Sigma-Aldrich®
4-Iodophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₆BIO₂
MDL Number: MFCD09953499
CAS číslo: 73852-88-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR