Tisk…

Magnesium aluminate, nanopowder <50 nm (BET) (Spinel), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dialuminium magnesium tetraoxide, Magnesium aluminum oxide, Aluminum magnesium oxide

M-Clarity™ quality level = MQ100

677396-5G
SIAL677396-5GEA 2850 CZK
SIAL677396-5G
Magnesium aluminate, nanopowder <50 nm (BET) (Spinel), Sigma-Aldrich®
Magnesium aluminate
Vzorec: Al₂MgO₄
MDL Number: MFCD00049475
CAS číslo: 12068-51-8
EINECS: 235-100-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR