Tisk…

Cyclopentyl methyl ether (CPME), Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

675970-100ML 675970-1L 675970-2L
SIAL675970-100MLEA 2150 CZK
SIAL675970-100ML SIAL675970-1L SIAL675970-2L
Cyclopentyl methyl ether (CPME), Sigma-Aldrich®
Cyclopentyl methyl ether (CPME)
Vzorec: C₆H₁₂O
Hustota: 0,86 g/cm³ (20 °C)
MDL Number: MFCD08276401
CAS číslo: 5614-37-9
UN: 3271
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR