Tisk…

(R)-(-)-Nipecotic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

656356-10G
SIAL656356-10GEA 8010 CZK
SIAL656356-10G
(R)-(-)-Nipecotic acid, Sigma-Aldrich®
(R)-(-)-Nipecotic acid
Vzorec: C₆H₁₁NO₂
MDL Number: MFCD01630787
CAS číslo: 25137-00-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR