Tisk…

3-(4-Chlorophenyl)propionic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-(4-Chlorophenyl)propanoic acid

M-Clarity™ quality level = MQ100

656151-5G
SIAL656151-5GEA 3990 CZK
SIAL656151-5G
3-(4-Chlorophenyl)propionic acid, Sigma-Aldrich®
3-(4-Chlorophenyl)propionic acid
Vzorec: C₉H₉ClO₂
MW: 184.62
MDL Number: MFCD00016555
CAS číslo: 2019-34-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR