Tisk…

2-Pyrimidinecarbonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Cyanopyrimidine

M-Clarity™ quality level = MQ100

646830-25G 646830-5G
SIAL646830-25GEA 3090 CZK
SIAL646830-25G SIAL646830-5G
2-Pyrimidinecarbonitrile, Sigma-Aldrich®
2-Pyrimidinecarbonitrile
Vzorec: C₅H₃N₃
MDL Number: MFCD00160513
CAS číslo: 14080-23-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR